tert-Butyl-4-[4-(tert-butyl)anilino]tetrahydro-1(2H)-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

042867-500MG 042867-5G
101857-946EA 278.46 USD
101857-946 101857-950
tert-Butyl-4-[4-(tert-butyl)anilino]tetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-4-[4-(tert-butyl)anilino]tetrahydro-1(2H)-pyridinecarboxylate

Hazard: Irritant

Formula: C₂₀H₃₂N₂O₂
MW: 332.49 g/mol
Melting Pt: 115…116 °C
MDL Number: MFCD06659533
CAS Number: 885949-49-5

Order Now

Specification Test Results

Melting point 115-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR