4'-Fluoro-2-hydroxyacetophenone ≥95%

Supplier: Matrix Scientific

042861-500MG 042861-5G
101857-910EA 15.38 USD
101857-910 101857-914
4'-Fluoro-2-hydroxyacetophenone ≥95%
4'-Fluoro-2-hydroxyacetophenone

Hazard: Irritant

Formula: C₈H₇FO₂
Melting Pt: 113…117 °C
MDL Number: MFCD02679035
CAS Number: 403-31-6

Order Now

Specification Test Results

Melting point 113-117°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR