4-Fluoro-4'-methoxy[1,1'-biphenyl]-3-carbonitrile ≥95%

Supplier: Matrix Scientific

042857-500MG 042857-5G
101857-890EA 307.69 USD
101857-890 101857-894
4-Fluoro-4'-methoxy[1,1'-biphenyl]-3-carbonitrile ≥95%
4-Fluoro-4'-methoxy[1,1'-biphenyl]-3-carbonitrile

Hazard: Irritant

Formula: C₁₄H₁₀FNO
Melting Pt: 94…96 °C
MDL Number: MFCD08056603
CAS Number: 885950-44-7

Order Now

Specification Test Results

Melting point 94-96°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR