3-Fluoro-4'-methoxy[1,1'-biphenyl]-4-carbonitrile ≥95%

Supplier: Matrix Scientific

042853-500MG 042853-5G
101857-866EA 556.92 USD
101857-866 101857-870
3-Fluoro-4'-methoxy[1,1'-biphenyl]-4-carbonitrile ≥95%
3-Fluoro-4'-methoxy[1,1'-biphenyl]-4-carbonitrile

Hazard: Irritant

Formula: C₁₄H₁₀FNO
MW: 227.24 g/mol
Melting Pt: 151…152 °C
MDL Number: MFCD06797371
CAS Number: 123864-93-7

Order Now

Specification Test Results

Melting point 151-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR