4-Chloro-5-hydroxy-3-(4-methoxyphenoxy)-2(5H)-furanone ≥95%

Supplier: Matrix Scientific

042850-500MG 042850-5G
101857-854EA 181.54 USD
101857-854 101857-858
4-Chloro-5-hydroxy-3-(4-methoxyphenoxy)-2(5H)-furanone ≥95%
4-Chloro-5-hydroxy-3-(4-methoxyphenoxy)-2(5H)-furanone

Hazard: Irritant

Formula: C₁₁H₉ClO₅
Melting Pt: 103…106 °C
MDL Number: MFCD06659532
CAS Number: 42190-28-3

Order Now

Specification Test Results

Melting point 103-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR