3-Amino-5-(trifluoromethyl)-1H-indazole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Trifluoromethyl)-1H-indazol-3-amine

042848-500MG 042848-5G
101857-842EA 278.46 USD
101857-842 101857-846
3-Amino-5-(trifluoromethyl)-1H-indazole ≥95%
3-Amino-5-(trifluoromethyl)-1H-indazole

Hazard: Irritant

Formula: C₈H₆F₃N₃
MW: 201.15 g/mol
Melting Pt: 114-118°C
MDL Number: MFCD01719740
CAS Number: 2250-53-5

Order Now

Specification Test Results

Melting point 114-118°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR