Methyl-4-amino-3-bromo-5-nitrobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-amino-3-bromo-5-nitrobenzoate

042845-500MG 042845-5G
101857-830EA 278.46 USD
101857-830 101857-834
Methyl-4-amino-3-bromo-5-nitrobenzoate ≥95%
Methyl 4-amino-3-bromo-5-nitrobenzoate

Hazard: Irritant

Formula: C₈H₇BrN₂O₄
Melting Pt: 108…110 °C
MDL Number: MFCD08443938
CAS Number: 105655-17-2

Order Now

Specification Test Results

Melting point 108-110°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR