Methyl-2-amino-5-(4-fluorophenyl)-1,3-thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

042840-500MG 042840-1G 042840-5G
101857-802EA 181.54 USD
101857-802 101857-804 101857-806
Methyl-2-amino-5-(4-fluorophenyl)-1,3-thiazole-4-carboxylate ≥95%
Methyl-2-amino-5-(4-fluorophenyl)-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₁H₉FN₂O₂S
MW: 252.27 g/mol
Melting Pt: 202 °C
MDL Number: MFCD06200913
CAS Number: 886361-30-4

Order Now

Specification Test Results

Melting point 202°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR