2-[(2-Ethoxy-2-oxoethyl)sulfanyl]benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[(2-Ethoxy-2-oxoethyl)thio]benzoic acid

042837-500MG 042837-1G 042837-5G
101857-784EA 181.54 USD
101857-784 101857-786 101857-788
2-[(2-Ethoxy-2-oxoethyl)sulfanyl]benzenecarboxylic acid ≥95%
2-[(2-Ethoxy-2-oxoethyl)sulfanyl]benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₂O₄S
Melting Pt: 113…114 °C
MDL Number: MFCD02364338
CAS Number: 18926-41-5

Order Now

Specification Test Results

Melting point 113-114°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR