Methyl-2-amino-5-(1-ethylpropyl)-1,3-thiazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

042836-500MG 042836-1G 042836-5G
101857-778EA 181.54 USD
101857-778 101857-780 101857-782
Methyl-2-amino-5-(1-ethylpropyl)-1,3-thiazole-4-carboxylate ≥95%
Methyl-2-amino-5-(1-ethylpropyl)-1,3-thiazole-4-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₆N₂O₂S
MW: 228.32 g/mol
Melting Pt: 81 °C
MDL Number: MFCD06200912
CAS Number: 886361-28-0

Order Now

Specification Test Results

Melting point 81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR