N'-Hydroxy-4-(1,3-oxazol-5-yl)benzenecarboximidamide ≥95%

Supplier: Matrix Scientific

042832-500MG 042832-1G 042832-5G
101857-766EA 556.92 USD
101857-766 101857-768 101857-770
N'-Hydroxy-4-(1,3-oxazol-5-yl)benzenecarboximidamide ≥95%
N'-Hydroxy-4-(1,3-oxazol-5-yl)benzenecarboximidamide

Hazard: Irritant

Formula: C₁₀H₉N₃O₂
Melting Pt: 203…205 °C
MDL Number: MFCD08276945
CAS Number: 860649-01-0

Order Now

Specification Test Results

Melting point 203-205°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR