3-[(2-Chloro-1,3-thiazol-5-yl)methoxy]-N'-hydroxybenzenecarboximidamide ≥95%
Supplier: Matrix Scientific
042829-500MG
042829-1G
042829-5G
101857-760EA
556.92
USD
101857-760
101857-762
101857-764
3-[(2-Chloro-1,3-thiazol-5-yl)methoxy]-N'-hydroxybenzenecarboximidamide ≥95%
3-[(2-Chloro-1,3-thiazol-5-yl)methoxy]-N'-hydroxybenzenecarboximidamide
Hazard: Irritant
Formula:
C₁₁H₁₀ClN₃O₂S Melting Pt: 153…155 °C |
MDL Number:
MFCD07698576 CAS Number: 885950-42-5 |
Specification Test Results
Melting point | 153-155°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...