Ethyl-3,5-dimethyl-1H-pyrazole-4-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 3,5-dimethyl-1H-pyrazole-4-carboxylate
042828-1G
042828-5G
101857-756EA
76.92
USD
101857-756
101857-758
Ethyl-3,5-dimethyl-1H-pyrazole-4-carboxylate ≥95%
Ethyl-3,5-dimethyl-1H-pyrazole-4-carboxylate
Hazard: Irritant
Formula:
C₈H₁₂N₂O₂ MW: 168.2 g/mol Melting Pt: 92…94 °C |
MDL Number:
MFCD00159638 CAS Number: 35691-93-1 |
Specification Test Results
Melting point | 92-94°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...