3-(1,3-Oxazol-5-yl)aniline ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(5-Oxazolyl)aniline

042827-500MG 042827-1G 042827-5G
101857-748EA 38.46 USD
101857-748 101857-750 101857-752
3-(1,3-Oxazol-5-yl)aniline ≥95%
3-(1,3-Oxazol-5-yl)aniline

Hazard: Irritant

Formula: C₉H₈N₂O
Melting Pt: 89…91 °C
MDL Number: MFCD00052191
CAS Number: 157837-31-5

Order Now

Specification Test Results

Melting point 89-91°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR