4,5-Dibromo-N-(3-hydroxypropyl)-1H-pyrrole-2-carboxamide ≥95%

Supplier: Matrix Scientific

Synonyms: 4,5-Dibromo-2-[(3-hydroxyprop-1-yl)carbamoyl]-1H-pyrrole

042824-500MG 042824-1G 042824-5G
101857-736EA 246.15 USD
101857-736 101857-738 101857-740
4,5-Dibromo-N-(3-hydroxypropyl)-1H-pyrrole-2-carboxamide ≥95%
4,5-Dibromo-N-(3-hydroxypropyl)-1H-pyrrole-2-carboxamide

Hazard: Irritant

Formula: C₈H₁₀Br₂N₂O₂
MW: 326 g/mol
Melting Pt: 160…162 °C
MDL Number: MFCD06200910
CAS Number: 186956-98-9

Order Now

Specification Test Results

Melting point 160-162°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR