Methyl-4-(5-formyl-2-thienyl)benzoate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(5-Formyl-2-thienyl)benzoic acid methyl ester, Methyl-4-(5-formyl-2-thienyl)benzenecarboxylate

042823-500MG 042823-1G 042823-5G
101857-730EA 556.92 USD
101857-730 101857-732 101857-734
Methyl-4-(5-formyl-2-thienyl)benzoate ≥95%
Methyl-4-(5-formyl-2-thienyl)benzoate

Hazard: Irritant

Formula: C₁₃H₁₀O₃S
MW: 246.29 g/mol
Melting Pt: 146…148 °C
MDL Number: MFCD06200909
CAS Number: 639816-37-8

Order Now

Specification Test Results

Melting point 146-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR