5-(4-Methoxyphenyl)-2-thiophenecarbaldehyde ≥95%
Supplier: Matrix Scientific
042822-500MG
042822-1G
042822-5G
101857-724EA
556.92
USD
101857-724
101857-726
101857-728
5-(4-Methoxyphenyl)-2-thiophenecarbaldehyde ≥95%
5-(4-Methoxyphenyl)-2-thiophenecarbaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₀O₂S Melting Pt: 117…119 °C |
MDL Number:
MFCD00663639 CAS Number: 38401-67-1 |
Specification Test Results
Melting point | 117-119°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...