Methyl-4,5-dibromo-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

042820-500MG 042820-1G 042820-5G
101857-712EA 180 USD
101857-712 101857-714 101857-716
Methyl-4,5-dibromo-1H-pyrrole-2-carboxylate ≥95%
Methyl-4,5-dibromo-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₆H₅Br₂NO₂
Melting Pt: 163…165 °C
MDL Number: MFCD06200907
CAS Number: 937-16-6

Order Now

Specification Test Results

Melting point 163-165°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR