4,5-Dibromo-1H-pyrrole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

042819-500MG 042819-1G 042819-5G
101857-706EA 230.77 USD
101857-706 101857-708 101857-710
4,5-Dibromo-1H-pyrrole-2-carboxylic acid ≥95%
4,5-Dibromo-1H-pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₅H₃Br₂NO₂
Melting Pt: >300 °C
MDL Number: MFCD01348229
CAS Number: 34649-21-3

Order Now

Specification Test Results

Melting point >300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR