tert-Butyl-2-(hydroxymethyl)-7,8-dihydro-4H-pyrazolo[1,5-a][1,4]diazepine-5(6H)-carboxylate ≥95%
Supplier: Matrix Scientific
042817-500MG
042817-1G
101857-696EA
710.77
USD
101857-696
101857-698
tert-Butyl-2-(hydroxymethyl)-7,8-dihydro-4H-pyrazolo[1,5-a][1,4]diazepine-5(6H)-carboxylate ≥95%
tert-Butyl-2-(hydroxymethyl)-7,8-dihydro-4H-pyrazolo[1,5-a][1,4]diazepine-5(6H)-carboxylate
Hazard: Irritant
Formula:
C₁₃H₂₁N₃O₃ MW: 267.33 g/mol Melting Pt: 55…57 °C |
MDL Number:
MFCD12025827 CAS Number: 1135282-77-7 |
Specification Test Results
Melting point | 55-57°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...