Pentafluorophenyl phenylmethanesulfonate ≥95%
Supplier: Matrix Scientific
042814-500MG
042814-1G
042814-5G
101857-680EA
375.38
USD
101857-680
101857-682
101857-684
Pentafluorophenyl phenylmethanesulfonate ≥95%
Pentafluorophenyl phenylmethanesulphonate
Hazard: Irritant
Formula:
C₁₃H₇F₅O₃S Melting Pt: 91…93 °C |
MDL Number:
MFCD05975135 CAS Number: 864450-16-8 |
Specification Test Results
Melting point | 91-93°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...