Pentafluorophenyl 4-tert-butylbenzenesulfonate ≥95%

Supplier: Matrix Scientific

Synonyms: 2,3,4,5,6-pentafluorophenyl 4-(tert-butyl)benzenesulphonate

042809-500MG 042809-1G 042809-5G
101857-650EA 375.38 USD
101857-650 101857-652 101857-654
Pentafluorophenyl 4-tert-butylbenzenesulfonate ≥95%
Pentafluorophenyl-4-tert-butylbenzenesulphonate

Hazard: Irritant

Formula: C₁₆H₁₃F₅O₃S
Melting Pt: 93-95°C
MDL Number: MFCD05975131
CAS Number: 885950-38-9

Order Now

Specification Test Results

Melting point 93-95°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR