Pentafluorophenyl 4-methoxybenzenesulfonate ≥95%

Supplier: Matrix Scientific

042805-500MG 042805-1G 042805-5G
101857-626EA 375.38 USD
101857-626 101857-628 101857-630
Pentafluorophenyl 4-methoxybenzenesulfonate ≥95%
Pentafluorophenyl 4-methoxybenzenesulphonate

Hazard: Irritant

Formula: C₁₃H₇F₅O₄S
Melting Pt: 58-60°C
MDL Number: MFCD05975127
CAS Number: 663175-94-8

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR