Pentafluorophenyl 4-methylbenzenesulfonate ≥95%
Supplier: Matrix Scientific
Synonyms:
Pentafluorophenyl toluene-4-sulphonate
042804-500MG
042804-1G
042804-5G
101857-620EA
375.38
USD
101857-620
101857-622
101857-624
Pentafluorophenyl 4-methylbenzenesulfonate ≥95%
Pentafluorophenyl 4-methylbenzenesulphonate
Hazard: Irritant
Formula:
C₁₃H₇F₅O₃S Melting Pt: 65…67 °C |
MDL Number:
MFCD00060056 CAS Number: 2069-36-5 |
Specification Test Results
Melting point | 65-67°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...