Pentafluorophenyl 2-nitrobenzenesulfonate ≥95%
Supplier: Matrix Scientific
042799-500MG
042799-1G
042799-5G
101857-590EA
375.38
USD
101857-590
101857-592
101857-594
Pentafluorophenyl 2-nitrobenzenesulfonate ≥95%
Pentafluorophenyl 2-nitrobenzenesulphonate
Hazard: Irritant
Formula:
C₁₂H₄F₅NO₅S Melting Pt: 127…128 °C |
MDL Number:
MFCD05975122 CAS Number: 885950-33-4 |
Specification Test Results
Melting point | 127-128°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...