Pentafluorophenyl 3-nitrobenzenesulfonate ≥95%
Supplier: Matrix Scientific
042798-500MG
042798-1G
042798-5G
101857-584EA
375.38
USD
101857-584
101857-586
101857-588
Pentafluorophenyl 3-nitrobenzenesulfonate ≥95%
Pentafluorophenyl 3-nitrobenzenesulphonate
Hazard: Irritant
Formula:
C₁₂H₄F₅NO₅S Melting Pt: 67-68°C |
MDL Number:
MFCD05975121 CAS Number: 663175-93-7 |
Specification Test Results
Melting point | 67-68°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...