Pentafluorophenyl 4-nitrobenzenesulfonate ≥95%

Supplier: Matrix Scientific

Synonyms: Pentafluorophenyl 4-Nitrobenzenesulphonate

042797-500MG 042797-1G 042797-5G
101857-578EA 375.38 USD
101857-578 101857-580 101857-582
Pentafluorophenyl 4-nitrobenzenesulfonate ≥95%
Pentafluorophenyl 4-nitrobenzenesulphonate

Hazard: Irritant

Formula: C₁₂H₄F₅NO₅S
MW: 369.2209 g/mol
Melting Pt: 109-111°C
MDL Number: MFCD05975120
CAS Number: 244633-31-6

Order Now

Specification Test Results

Melting point 109-111°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR