Methyl-2-(bromomethyl)-3-nitrobenzoate ≥95%

Supplier: Matrix Scientific

042794-1G 042794-5G
101857-564EA 29.23 USD
101857-564 101857-566
Methyl-2-(bromomethyl)-3-nitrobenzoate ≥95%
Methyl 2-(bromomethyl)-3-nitrobenzoate

Hazard: Irritant

Formula: C₉H₈BrNO₄
Melting Pt: 72…74 °C
MDL Number: MFCD04114315
CAS Number: 98475-07-1

Order Now

Specification Test Results

Melting point 72-74°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR