2-Amino-3,6-dimethoxybenzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

042792-500MG 042792-1G 042792-5G
101857-552EA 747.69 USD
101857-552 101857-554 101857-556
2-Amino-3,6-dimethoxybenzenecarbaldehyde ≥95%
2-Amino-3,6-dimethoxybenzenecarbaldehyde

Hazard: Irritant

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Melting Pt: 66…68 °C
MDL Number: MFCD03848397
CAS Number: 126522-16-5

Order Now

Specification Test Results

Melting point 66-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR