2-(1,2-Benzisoxazol-3-yl)acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1,2-Benzisoxazole-3-acetic Acid

042791-500MG 042791-1G 042791-5G
101857-546EA 181.54 USD
101857-546 101857-548 101857-550
2-(1,2-Benzisoxazol-3-yl)acetic acid ≥95%
2-(1,2-Benzisoxazol-3-yl)acetic acid

Hazard: Irritant

Formula: C₉H₇NO₃
MW: 177.1568 g/mol
Melting Pt: 130-133°C
MDL Number: MFCD02180388
CAS Number: 4865-84-3

Order Now

Specification Test Results

Melting point 130-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR