6-Aminonicotinohydrazide ≥95%

Supplier: Matrix Scientific

042785-500MG 042785-1G 042785-5G
101857-520EA 556.92 USD
101857-520 101857-522 101857-524
6-Aminonicotinohydrazide ≥95%
6-Aminonicotinohydrazide

Hazard: Irritant

Formula: C₆H₈N₄O
Melting Pt: 228…232 °C
MDL Number: MFCD03844845
CAS Number: 42596-56-5

Order Now

Specification Test Results

Melting point 228-232°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR