2-[2-(Hydroxymethyl)phenoxy]benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[2-(Hydroxymethyl)phenoxy]benzonitrile

042783-500MG 042783-1G 042783-5G
101857-508EA 747.69 USD
101857-508 101857-510 101857-512
2-[2-(Hydroxymethyl)phenoxy]benzenecarbonitrile ≥95%
2-[2-(Hydroxymethyl)phenoxy]benzenecarbonitrile

Hazard: Irritant

Formula: C₁₄H₁₁NO₂
MW: 225.25 g/mol
Melting Pt: 54…57 °C
MDL Number: MFCD03844844
CAS Number: 59167-65-6

Order Now

Specification Test Results

Melting point 54-57°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR