Ethyl-2-oxo-2-(1H-pyrrolo[2,3-b]pyridin-3-yl)-acetate ≥95%
Supplier: Matrix Scientific
042781-500MG
042781-1G
101857-500EA
195.38
USD
101857-500
101857-502
Ethyl-2-oxo-2-(1H-pyrrolo[2,3-b]pyridin-3-yl)-acetate ≥95%
Ethyl-2-oxo-2-(1H-pyrrolo[2,3-b]pyridin-3-yl)-acetate
Hazard: Irritant
Formula:
C₁₁H₁₀N₂O₃ Melting Pt: 160…162 °C |
MDL Number:
MFCD05256119 CAS Number: 626604-80-6 |
Specification Test Results
Melting point | 160-162°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...