Methyl-3-iodo-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

042779-500MG 042779-1G 042779-5G
101857-488EA 278.46 USD
101857-488 101857-490 101857-492
Methyl-3-iodo-1H-indole-2-carboxylate ≥95%
Methyl-3-iodo-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₈INO₂
MW: 301.09 g/mol
Melting Pt: 157…159 °C
MDL Number: MFCD04124152
CAS Number: 534595-85-2

Order Now

Specification Test Results

Melting point 157-159°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR