3-Methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine ≥95%

Supplier: Matrix Scientific

042774-500MG 042774-1G 042774-5G
101857-466EA 278.46 USD
101857-466 101857-468 101857-470
3-Methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine ≥95%
3-Methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₁₁H₁₀F₃N₃
Melting Pt: 110…113 °C
MDL Number: MFCD03791147
CAS Number: 851975-08-1

Order Now

Specification Test Results

Melting point 110-113°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR