1-(4-Fluorophenyl)-3-methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine ≥95%

Supplier: Matrix Scientific

042772-500MG 042772-1G 042772-5G
101857-454EA 278.46 USD
101857-454 101857-456 101857-458
1-(4-Fluorophenyl)-3-methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine ≥95%
1-(4-Fluorophenyl)-3-methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₁₇H₁₃F₄N₃
Melting Pt: 109…112 °C
MDL Number: MFCD03791145
CAS Number: 446275-99-6

Order Now

Specification Test Results

Melting point 109-112°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR