3-[4-(2-Fluorobenzoyl)-1-methyl-1H-pyrrol-2-yl]-2-[3-(trifluoromethyl)phenyl]-2-propenenitrile ≥95%

Supplier: Matrix Scientific

042771-500MG 042771-1G 042771-5G
101857-448EA 375.38 USD
101857-448 101857-450 101857-452
3-[4-(2-Fluorobenzoyl)-1-methyl-1H-pyrrol-2-yl]-2-[3-(trifluoromethyl)phenyl]-2-propenenitrile ≥95%
3-[4-(2-Fluorobenzoyl)-1-methyl-1H-pyrrol-2-yl]-2-[3-(trifluoromethyl)phenyl]-2-propenenitrile

Hazard: Irritant

Formula: C₂₂H₁₄F₄N₂O
MW: 398.36 g/mol
Melting Pt: 244…246 °C
MDL Number: MFCD05021838
CAS Number: 885951-46-2

Order Now

Specification Test Results

Melting point 244-246°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR