Ethyl-4,6-bis(4-chlorophenyl)-2-oxo-3-cyclohexene-1-carboxylate ≥95%

Supplier: Matrix Scientific

042769-500MG 042769-1G 042769-5G
101857-436EA 278.46 USD
101857-436 101857-438 101857-440
Ethyl-4,6-bis(4-chlorophenyl)-2-oxo-3-cyclohexene-1-carboxylate ≥95%
Ethyl-4,6-bis(4-chlorophenyl)-2-oxo-3-cyclohexene-1-carboxylate

Hazard: Irritant

Formula: C₂₁H₁₈Cl₂O₃
Melting Pt: 124…126 °C
MDL Number: MFCD03425713
CAS Number: 26379-96-4

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR