Ethyl-3-(4-benzoyl-1-methyl-1H-pyrrol-2-yl)-acrylate ≥95%
Supplier: Matrix Scientific
042768-500MG
042768-1G
042768-5G
101857-430EA
375.38
USD
101857-430
101857-432
101857-434
Ethyl-3-(4-benzoyl-1-methyl-1H-pyrrol-2-yl)-acrylate ≥95%
Ethyl-3-(4-benzoyl-1-methyl-1H-pyrrol-2-yl)-acrylate
Hazard: Irritant
Formula:
C₁₇H₁₇NO₃ Melting Pt: 96…98 °C |
MDL Number:
MFCD03425712 CAS Number: 128843-39-0 |
Specification Test Results
Melting point | 96-98°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...