7-Hydroxy-6-propyl-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-3-propyl-1-azatricyclo[7.3.1.05,13]trideca-3,5(13),6,8-tetraen-2-one

042767-500MG 042767-1G 042767-5G
101857-424EA 181.54 USD
101857-424 101857-426 101857-428
7-Hydroxy-6-propyl-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one ≥95%
7-Hydroxy-6-propyl-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one

Hazard: Irritant

Formula: C₁₅H₁₇NO₂
MW: 243.31 g/mol
Melting Pt: 170…172 °C
MDL Number: MFCD03791142
CAS Number: 57625-51-1

Order Now

Specification Test Results

Melting point 170-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR