Ethyl-4-(2-methoxyphenyl)-2-oxo-6-(3-thienyl)-3-cyclohexene-1-carboxylate ≥95%

Supplier: Matrix Scientific

042765-500MG 042765-1G 042765-5G
101857-412EA 278.46 USD
101857-412 101857-414 101857-416
Ethyl-4-(2-methoxyphenyl)-2-oxo-6-(3-thienyl)-3-cyclohexene-1-carboxylate ≥95%
Ethyl-4-(2-methoxyphenyl)-2-oxo-6-(3-thienyl)-3-cyclohexene-1-carboxylate

Hazard: Irritant

Formula: C₂₀H₂₀O₄S
Melting Pt: 145…147 °C
MDL Number: MFCD04113617
CAS Number: 446275-96-3

Order Now

Specification Test Results

Melting point 145-147°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR