Ethyl-3-[4-(2,4-dichlorobenzoyl)-1-methyl-1H-pyrrol-2-yl]acrylate ≥95%

Supplier: Matrix Scientific

042764-500MG 042764-1G 042764-5G
101857-406EA 375.38 USD
101857-406 101857-408 101857-410
Ethyl-3-[4-(2,4-dichlorobenzoyl)-1-methyl-1H-pyrrol-2-yl]acrylate ≥95%
Ethyl-3-[4-(2,4-dichlorobenzoyl)-1-methyl-1H-pyrrol-2-yl]acrylate

Hazard: Irritant

Formula: C₁₇H₁₅Cl₂NO₃
Melting Pt: 86…89 °C
MDL Number: MFCD02571785
CAS Number: 400077-69-2

Order Now

Specification Test Results

Melting point 86-89°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR