(4-Nitrophenyl)(piperazino)methanone ≥95%
Supplier: Matrix Scientific
Synonyms:
(4-Nitrophenyl)(piperazin-1-yl)methanone, 1-(4-Nitrobenzoyl)piperazine
042758-500MG
042758-1G
042758-5G
101857-376EA
181.54
USD
101857-376
101857-378
101857-380
(4-Nitrophenyl)(piperazino)methanone ≥95%
(4-Nitrophenyl)(piperazino)methanone
Hazard: Irritant
Formula:
C₁₁H₁₃N₃O₃ MW: 235.24 g/mol Melting Pt: 158…161 °C |
MDL Number:
MFCD00661867 CAS Number: 72141-41-4 |
Specification Test Results
Melting point | 158-161°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...