4-(Cyclohexylcarbonyl)-1H-pyrrole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

042757-500MG 042757-1G
101857-372EA 921.54 USD
101857-372 101857-374
4-(Cyclohexylcarbonyl)-1H-pyrrole-2-carboxylic acid ≥95%
4-(Cyclohexylcarbonyl)-1H-pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₅NO₃
Melting Pt: 220…222 °C
MDL Number: MFCD02571980
CAS Number: 477848-96-7

Order Now

Specification Test Results

Melting point 220-222°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR