2-Cyclohexyl-3-(4-methoxyphenyl)-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid ≥95%

Supplier: Matrix Scientific

042756-500MG 042756-1G 042756-5G
101857-366EA 230.77 USD
101857-366 101857-368 101857-370
2-Cyclohexyl-3-(4-methoxyphenyl)-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid ≥95%
2-Cyclohexyl-3-(4-methoxyphenyl)-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid

Hazard: Irritant

Formula: C₂₃H₂₅NO₄
Melting Pt: 171…172 °C
MDL Number: MFCD03001220
CAS Number: 385383-39-1

Order Now

Specification Test Results

Melting point 171-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR