Ethyl-4-(4-ethoxyphenyl)-6-methyl-2-thioxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%

Supplier: Matrix Scientific

042752-500MG 042752-1G 042752-5G
101857-344EA 181.54 USD
101857-344 101857-346 101857-348
Ethyl-4-(4-ethoxyphenyl)-6-methyl-2-thioxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate ≥95%
Ethyl-4-(4-ethoxyphenyl)-6-methyl-2-thioxo-1,2,3,4-tetrahydro-5-pyrimidinecarboxylate

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₃S
Melting Pt: 156…158 °C
MDL Number: MFCD00409203
CAS Number: 293764-13-3

Order Now

Specification Test Results

Melting point 156-158°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR