Methyl 4-cyclohexanecarbonyl-1H-pyrrole-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl-4-(cyclohexylcarbonyl)-1H-pyrrole-2-carboxylate
042750-500MG
042750-1G
042750-5G
101857-332EA
747.69
USD
101857-332
101857-334
101857-336
Methyl 4-cyclohexanecarbonyl-1H-pyrrole-2-carboxylate ≥95%
Methyl 4-cyclohexanecarbonyl-1H-pyrrole-2-carboxylate
Hazard: Irritant
Formula:
C₁₃H₁₇NO₃ MW: 235.279 g/mol Melting Pt: 168…170 °C |
MDL Number:
MFCD02571975 CAS Number: 477848-90-1 |
Specification Test Results
Melting point | 168-170°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...