6-(4-Methylphenyl)-2-oxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid ≥95%
Supplier: Matrix Scientific
042745-500MG
042745-1G
042745-5G
101857-302EA
278.46
USD
101857-302
101857-304
101857-306
6-(4-Methylphenyl)-2-oxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid ≥95%
6-(4-Methylphenyl)-2-oxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O₃ Melting Pt: 248…250 °C |
MDL Number:
MFCD03425706 CAS Number: 446275-91-8 |
Specification Test Results
Melting point | 248-250°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...