5-Chloro-2-(2-propynyloxy)benzenecarbaldehydeoxime ≥95%

Supplier: Matrix Scientific

042744-500MG 042744-1G 042744-5G
101857-296EA 375.38 USD
101857-296 101857-298 101857-300
5-Chloro-2-(2-propynyloxy)benzenecarbaldehydeoxime ≥95%
5-Chloro-2-(2-propynyloxy)benzenecarbaldehydeoxime

Hazard: Irritant

Formula: C₁₀H₈ClNO₂
Melting Pt: 120…123 °C
MDL Number: MFCD03001218
CAS Number: 400751-52-2

Order Now

Specification Test Results

Melting point 120-123°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR