[5-Chloro-2-(2-propynyloxy)phenyl]methanol ≥95%
Supplier: Matrix Scientific
042741-500MG
042741-1G
042741-5G
101857-278EA
375.38
USD
101857-278
101857-280
101857-282
[5-Chloro-2-(2-propynyloxy)phenyl]methanol ≥95%
[5-Chloro-2-(2-propynyloxy)phenyl]methanol
Hazard: Irritant
Formula:
C₁₀H₉ClO₂ Melting Pt: 55…58 °C |
MDL Number:
MFCD03001217 CAS Number: 400751-45-3 |
Specification Test Results
Melting point | 55-58°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...